Learn More
ATCC Human herpesvirus 5; Human cytomegalovirus; Strain: AD-169
Supplier:  ATCC VR538
Encompass
Human herpesvirus 5 strain AD-169 is propagated in MRC-5 cells (ATCC CCL-171). This strain was isolated from the adenoid tissue of a 7-year-old female. This product has applications in virucide testing.
Catalog No. 50-238-2206