Learn More
ATCC Human herpesvirus 5; Human cytomegalovirus; Strain: Towne
Supplier:  ATCC VR977
Encompass
Human herpesvirus 5 strain Towne is propagated in MRC-5 cells (ATCC CCL-171). This strain was isolated from the urine of a human male with congenital cytomegalic inclusion disease. This product has applications in infectious disease research.
Catalog No. 50-238-2156