Learn More
ATCC Human herpesvirus 5; Strain: Merlin; Human cytomegalovirus
Supplier:  ATCC VR1590
Encompass
Human herpesvirus 5 strain Merlin is propagated in MRC-5 cells (ATCC CCL-171). This strain was isolated from the urine of a congenitally infected infant and was deposited by the Cardiff Public Health Laboratory. It has applications in infectious disease and sexually transmitted disease research.
Catalog No. 50-238-3381