Learn More
BIOHIPPO Human HGFR Microsample
SDP
Supplier:  BIOHIPPO BHE15211641
Encompass
Human HGFR Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of HGFR in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 125-8000 pg/mL with a sensitivity of 47 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human HGFR Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human HGFR Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human HGFR biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15458