Learn More
Sino Biological Recombinant Anti-15-PGDH Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 11205R027F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human HPGD / 15-PGDH (rh HPGD / 15-PGDH; Catalog#11205-H08E; NP_000851.2; Met1-Gln266) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-241-0174