Learn More
American Research Products Inc Human Hyaluronidase PH 20 SPA
Supplier:  American Research Products Inc CSBEL022471HU
Encompass
Human Hyaluronidase PH-20 (SPAM1) ELISA kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma urine tissue homogenates Method Sandwich
Catalog No. 50-003-27871