Learn More
Abcam Human ICAM1 ELISA Kit (CD54), 1 x 96 Tests
Supplier:  Abcam AB1744451X96TESTS
Encompass_Preferred
Human ICAM1 ELISA Kit (CD54) is a single-wash 90-min Simplestep used to quantify Human ICAM1 (CD54) with a sensitivity of 1.6 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Cited in over 25 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2178448