Learn More
Abcam Human IFN gamma ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1744431X96TESTS
Encompass_Preferred
Human IFN gamma (IFNG) ELISA kit is a single-wash, 90-minute SimpleStep ELISA a used to quantify human interferon-gamma (IFN-γ) with a sensitivity of 470 pg/mL. The assay employs a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric sandwich ELISA - 450 nm readout; compatible with any standard plate reader - Available in different formats to meet various throughput needs, including 384-well plates for higher throughput - Broad sample compatibility – suitable for serum, plasma (citrate, EDTA), and cell culture supernatants - Cited in over 25 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2575571