Learn More
TRIBIOSCIENCE INC Human IFN-Gamma ELISA Kit, 1 x 96-well plate
SDP
Supplier:  TRIBIOSCIENCE INC TBS3230
Encompass_Preferred
The Human IFN-Gamma ELISA Maxi is a solid phase ELISA designed to measure human IFN-Gamma levels in cell culture supernatants, serum, and plasma. The kit uses proprietary methods to combine samples and detections in one step, rather than traditional ELISA methods. The detection range is from 20 to 15000 pg/mL.
Catalog No. NC2886047