Learn More
Mabtech Inc Human IFN-gamma ELISpot PRO kit (ALP), strips, 2 plates
Supplier:  Mabtech Inc 34202AST2
Encompass
The ELISpotPRO kit is Mabtech's most innovative product featuring pre-coated plates and one-step detection. The kit enables enumeration of cells secreting human IFN-gamma and contains 96-well ELISpot strip plates (8 wells x 12 strips) pre-coated with monoclonal antibody, ALP-conjugated antibody for one-step detection and BCIP/NBT-plus substrate. A polyclonal activator for human T cells (mAb CD3-2) is supplied as a positive control for cytokine secretion.
Catalog No. NC0609319