Learn More
Raybiotech Inc Quantibody Human Immunoglobulin Isotyping Array Kit.
Supplier:  Raybiotech Inc QAHISO11
Encompass_Preferred
Detects 8 Human Immunoglobulin Sub-Types. Suitable for serum or plasma. Quantitative, Sandwich-based, Glass Slide Antibody Array. Simultaneous quantitative measurement of multiple proteins in a wide variety of sample types. Quantibody combines the high specificity and sensitivity of ELISA with the high throughput of the glass chip-based array. Less sample, more data: only 50 ul of sample is needed for quantification of up to 40 cytokines. Proteins detected: IgA, IgD, IgE, IgG1, IgG2, IgG3, IgG4, IgM
Catalog No. 50-195-0845