Learn More
Genetex Inc Rabbit anti-human IgD Monoclonal Antibody [MSVA-701R] HistoMAX™ (GTX04491) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04491
Encompass_Preferred
The Rabbit anti-human IgD antibody [MSVA-701R] HistoMAX™ (GTX04491) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human thymus tissue using GTX04491 Rabbit anti-human IgD antibody [MSVA-701R] HistoMAX. Only few cells show human IgD staining especially in the medulla.
Catalog No. NC3668410