Learn More
Immunology Consultants Human IgE ELISA Kit
SDP
Supplier:  Immunology Consultants E80E
Encompass_Preferred
The Human IgE test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring IgE in human biological samples.
Catalog No. 50-240-6542