Learn More
Genscript Corporation Human IgE RC7H mAb Mouse
Supplier:  Genscript Corporation V0530110
Encompass
IgE is a type of immunoglobulins produced by the immune system It is composed of two heavy chains and two light chains like Ig molecules IgE plays an essential role in various allergic reactions Its main function is immunity to parasites such as Schistosoma mansoni Trichinella spiralis and Fasciola hepatica It also plays an important role in type I hypersensitivity Total IgE is considered as a significant marker of allergic state
Catalog No. 50-000-59039