Learn More
Rockland Immunochemicals Human IgG F(c) Antibody 5 mg
SDP
Supplier:  Rockland Immunochemicals 6091103
Encompass_Preferred
Anti-Human IgG F(c) generated in goat detects Human F(c). A proteolytic fragment of immunoglobulin G (IgG) obtained by limited digestion with the enzyme papain under controlled conditions of temperature time and pH. Receptors bind the Fc portion of human IgG and often this fragment is removed from immunoglobulins to minimize receptor binding and lower background reactivity. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition. F(c) Antibody is ideal for investigators who routinely perform flow cytometry immunohistochemistry or IHC and other immunoassays.
Catalog No. 50-194-3887