Learn More
MOLECULAR DEPOT LLC Human IgG Powder - 100 mg
Supplier:  MOLECULAR DEPOT LLC C2010428
Encompass_Preferred
Human Gamma Globulins Solution is a lyophilized preparation of highly pure Human Gamma Globulins (IgG fraction containing IgG IgM IgA IgD and IgE) This product can be used to test Human Gamma Globulins interference in assays This product is for in vitro research use only This product is not for human use
Catalog No. 50-000-13376