Learn More
Sigma Aldrich Fine Chemicals Biosciences IgG1, Kappa from human myeloma plasma, MFCD00163924, 1mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences I51541MG
Encompass
Purified immunoglobulin, >95%, Synonym: Human IgG1-κ; IgG antibody subtype is the most abundant serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses.
Catalog No. NC1667749