Learn More
Abcam Human IL-11 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1895691X96TESTS
Encompass_Preferred
Human IL-11 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human IL-11 with a sensitivity of 12 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Cited in over 5 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1683699