Learn More
ACROBiosystems Human IL-12p70 ELISA Kit
Supplier: ACROBiosystems CRSB00996TESTS
HumanInterleukin-12p70 (IL-12p70) OneStep ELISA Kit is a one-step quick immunoassay designed to measure human IL-12p70 levels in cell culture supernates, serum, and plasma. This assay kit is a standard sandwich-ELISA format. It contains recombinant human IL-12p70 and a pair of antibodies against the recombinant factor, which are provided by ACROBiosystems. Results are obtained by four parameter logistic curve that were parallel to the standard curves obtained. The verification results indicate that this kit can be used for the quantitative determination of natural and recombinant human IL-12p70 concentrations. The specificity has been verified.