Learn More
Bon Opus Biosciences Recombinant Human IL-15RA & IL-15 fusion protein (Fc) - Source: Human 293 Cells
SDP
Supplier:  Bon Opus Biosciences BP20150UG
Encompass_Preferred
Recombinant Human Interleukin-15 Receptor Alpha & Interleukin-15 Fusion Protein is produced by our Mammalian expression system and the target gene encoding Ile31-Asp96&Asn49-Ser162(Asn120Asp) is expressed with a human IgG1 Fc tag at the C-terminus. Accession#Q13261 & P40933. Formulation: Lyophilized from a 0.2 um filtered solution of PBS, pH7.4. Normally 5% - 8% trehalose, mannitol and 0.01% Tween80 are added as protectants before lyophilization.
Catalog No. NC2733657