Learn More
BIOHIPPO Human IL-2sRa/CD25 Microsample
SDP
Supplier:  BIOHIPPO BHE15212097
Encompass
Human IL-2sRa/CD25 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL-2sRa/CD25 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 15 63-1000 pg/mL with a sensitivity of 6 5 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL-2sRa/CD25 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL-2sRa/CD25 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL-2sRa/CD25 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14259