Learn More
Raybiotech Inc HUMAN IL-8 ELISA
Supplier:  Raybiotech Inc ELHIL81
Encompass_Preferred
RayBio Human IL-8 (CXCL8) ELISA Kit for cell culture supernatants, plasma, and serum samples. Pre-coated 96-well microplate kit with additional reagents. Detection Range: 1 pg/ml - 600 pg/ml
Catalog No. 50-104-4002