Learn More
Sino Biological Recombinant Human IL-11 Protein
Supplier:  Sino Biological 12225HNCE1100
Encompass_Preferred
A DNA sequence encoding the mature form of human IL11 (P20809-1) (Pro22-Leu199) was expressed.
Catalog No. 50-299-3616