Learn More
Biomatik Corporation Human IL-17E/IL-25 (Interleukin 25) ELISA Kit
Supplier:  Biomatik Corporation EKF58819
Encompass_Preferred
Human IL-17E/IL-25 (Interleukin 25) ELISA Kit for detecting IL-17E; IL-25 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.625-1000pg/ml. Sensitivity: 9.375pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q9H293. Target Synonyms: IL-25; IL-17E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4478