Learn More
Sino Biological Human IL18 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 310203T32100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human IL-18 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-237-0839