Learn More
Sino Biological Human IL21R MAb 100ul (Sino Biological)
Supplier:  Sino Biological 11483MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL-21R (rh IL-21R; Catalog#11483-H08H; Q9HBE5-1; Met1-Pro236). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1652