Learn More
BIOHIPPO Human IL28B Microsample
SDP
Supplier:  BIOHIPPO BHE15212198
Encompass
Human IL28B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IL28B in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 7 82-500 pg/mL with a sensitivity of 3 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IL28B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IL28B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IL28B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14144