Learn More
Sino Biological Recombinant Human IL-5R alpha Protein (Membrane-bound, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10392H01H1100
Encompass_Preferred
A DNA sequence encoding the Human IL5RA (Q01344-1) (Asp21-Trp342) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-299-2682