Learn More
Sino Biological Human ILT5/LILRB3 Matched ELISA Antibody Pair Set 5 Plates
Supplier:  Sino Biological SEK119785
Encompass_Preferred
The Human LILRB3 ELISA Pair Set is for the quantitative determination of Human LILRB3.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human LILRB3 coated on a 96-well plate. Standards and samples are added to the wells, and any Human LILRB3 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human LILRB3 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human LILRB3 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86622