Learn More
Sino Biological Human Immunodeficiency Virus type 1 (HIV-1) gp120 / Glycoprotein 120 ELISA Pair Set 5 Plates
Supplier:  Sino Biological SEK112335
Encompass_Preferred
The Human Immunodeficiency Virus type 1 (HIV-1) gp120 / Glycoprotein 120 ELISA Pair Set is for the quantitative determination of Human Immunodeficiency Virus type 1 (HIV-1) gp120 / Glycoprotein 120.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human Immunodeficiency Virus type 1 (HIV-1) gp120 / Glycoprotein 120 coated on a 96-well plate. Standards and samples are added to the wells, and any Human Immunodeficiency Virus type 1 (HIV-1) gp120 / Glycoprotein 120 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human Immunodeficiency Virus type 1 (HIV-1) gp120 / Glycoprotein 120 monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human Immunodeficiency Virus type 1 (HIV-1)
Catalog No. 50-222-1497