Learn More
Biotium Human Immunoglobulin Gamma (IgG) Heavy Chain(IG217 + IG266), CF405S conjugate, 0.1mg/mL
Supplier:  Biotium BNC041262100
Encompass_Preferred
This MAb is specific to heavy chain of IgG and shows minimal cross-reaction with heavy chains of other immunoglobulins. It is reactive with all subclasses of Gamma heavy chain(IgG1, IgG2a, IgG2b, IgG3). The most common feature of plasmacytomas, and certain non-Hodgkin's lymphomas is the restricted expression of a single heavy chain class. Demonstration of clonality in lymphoid infiltrates indicates that the infiltrate is clonal and therefore malignant. Ab-3 is superb for staining of formalin-fixed tissues.
Catalog No. 50-006-24633