Learn More
ACROBiosystems ACROBiosystems Human Integrin alpha 5 beta 1 (ITGA5&ITGB1) Heterodimer Protein, His Tag&Tag Free
Supplier:  ACROBiosystems IT1H52W51MG
Encompass_Preferred
Recombinant Protein;1MG;Human ITGA5&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-H52W5) is expressed from human 293 cells (HEK293). It contains AA Phe 42 - Tyr 995 (ITGA5) & Gln 21 - Asp 728 (ITGB1) (Accession # P08648-1 (ITGA5) & P05556-1 (ITGB1)).;Human ITGA5&ITGB1 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGA5 and ITGB1, has a calculated MW of 110.7 kDa (ITGA5) and 83.7 kDa (ITGB1). Subunit ITGA5 is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB1 contains no tag but a basic tail at the C-terminus. The non-reducing (NR) protein migrates as 130-150 kDa (ITGA5) and 100-120 kDa (ITGB1) respectively due to glycosylation.;Immobilized Human Fibronectin at 2 μg/mL (100 μL/well) can bind Human ITGA5&ITGB1 Heterodimer Protein, His Tag&Tag Free (Cat. No. IT1-H52W5) with a linear range of 0.2-2 ng/mL (QC tested).
Catalog No. 50-210-6685