Learn More
ACROBiosystems ACROBiosystems Human Integrin alpha X beta 2 (ITGAX&ITGB2) Heterodimer Protein, His Tag&Tag Free
Supplier:  ACROBiosystems IT2H52W3100UG
Encompass_Preferred
Recombinant Protein;100UG;Human ITGAX&ITGB2 Heterodimer Protein, His Tag&Tag Free (IT2-H52W3) is expressed from human 293 cells (HEK293). It contains AA Phe 20 -Pro 1107 (ITGAX) & Gln 23 - Asn 700 (ITGB2) (Accession # P20702-1 (ITGAX) & P05107-1 (ITGB2)).;Human ITGAX&ITGB2 Heterodimer Protein, His Tag&Tag Free, produced by co-expression of ITGAX and ITGB2, has a calculated MW of 126.5 kDa (ITGAX) and 80.2 kDa (ITGB2). Subunit ITGAX is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB2 contains no tag but a basic tail at the C-terminus. The protein migrates as 145 kDa (ITGAX) and 90 kDa (ITGB2) under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-7324