Learn More
American Research Products Inc Human Interferon regulatory fa
Supplier:  American Research Products Inc CSBEL011822HU
Encompass
Human Interferon regulatory factor 7 (IRF7) ELISA kit from CUSABIO Detection Range 23 4 pg/mL-1500 pg/mL Sensitivity 5 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27092