Learn More
Biomatik Corporation Human Interleukin 20 (IL20) ELISA Kit
Supplier:  Biomatik Corporation EKU05282
Encompass_Preferred
Human Interleukin 20 (IL20) ELISA Kit for detecting IL20 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.62-1000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NYY1. Gene ID: 50604. Target Synonyms: IL10D; IL10-D; ZCYTO10; Cytokine Zcyto10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4852