Learn More
Sino Biological Human ITGA6 & ITGB4 Heterodimer Protein, Endotoxin-Free 1mg
Supplier:  Sino Biological CT069H2508HUE1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Ser 1011) of human ITGA6 isoform alpha-6X1B (P23229-3) was fused with a flag tag at the C-terminus, constructed the plasmid 1 ; A DNA sequence encoding the extracellular domain (Met 1-Ser 710) of human ITGB4 (CAA37656.1) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the heterodimer was purified
Catalog No. 50-003-31988