Learn More
ACROBiosystems Recombinant Protein;Human Integrin alpha V beta 6 (ITGAV&ITGB6) Heterodimer Protein;HEK293;100UG;IT6,Integrin alpha V beta 6,ITGAV&ITGB6,Integrin,ITGAV,ITGA,ITGB6,ITGB,ITGAVB6
Supplier:  ACROBiosystems IT6H52E1100UG
Encompass_Preferred
Recombinant Protein;Human Integrin alpha V beta 6 (ITGAV&ITGB6) Heterodimer Protein;100UG;Human ITGAV&ITGB6, His Tag (IT6-H52E1) is expressed from human 293 cells (HEK293). It contains AA Phe 31 - Val 992 (ITGAV) & Gly 22 - Asn 707 (ITGB6) (Accession # NP_002201 (ITGAV)& AAI21179 (ITGB6)).;Human ITGAV & ITGB6 Heterodimer Protein, produced by co-expression of ITGAV and ITGB6, has a calculated MW of 113 kDa (ITGAV) and 79.6 kDa (ITGB6). Subunit ITGAV is fused with an acidic tail at the C-terminus and followed by a polyhistidine tag and subunit ITGB6 contains no tag but a basic tail at the C-terminus. The predicted N-terminus is Phe 31 (ITGAV) & Gly 22 (ITGB6). The reducing (R) protein migrates as 110 kDa,130 kDa (ITGB6) and 145 kDa (ITGAV) respectively due to glycosylation.;Immobilized Human ITGAV&ITGB6, His Tag (Cat. No. IT6-H52E1) at 5 μg/mL (100 μL/well) can bind Biotinylated Human Latent TGF-Beta 1, His Tag, Avi Tag (Cat. No. TG1-H82Qb) with a linear range of 0.5-4 ng/mL (QC tested
Catalog No. 50-108-9645