Learn More
Sino Biological Anti-KRAS Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101667T3220
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human K-Ras, and purified by antigen affinity chromatography.
Catalog No. 50-237-0848