Learn More
ACROBiosystems ACROBiosystems Human KRAS (2-185,G12D) Protein, His Tag
Supplier:  ACROBiosystems KRSH51H41MG
Encompass_Preferred
Recombinant Protein;1MG;Human KRAS (2-185,G12D), His Tag (KRS-H51H4) is expressed from E.coli cells. It contains AA Thr 2 - Cys 185 (Accession # P01116-2 (G12D)).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 23.0 kDa.
Catalog No. 50-210-6809