Learn More
BIORTUS USA INC Human KRAS4B Protein, biotinylated/GMPPNP_loaded, 100UG
Supplier:  BIORTUS USA INC BP190730MB
Encompass_Preferred
Gene Name: KRAS. Uniprot: P01116. Construct: biotinylated-Avi-KRas4B(T2-M188 end) (G12D),GMPPNP loaded. Expression Host: E. coli. Purity: 90%. 1 each contains 100UG.
Catalog No. NC2953311