Learn More
Raybiotech Inc L-Series Human Antibody Array 493 Membrane Kit.
Supplier:  Raybiotech Inc AAHBLM2A4
Encompass_Preferred
Detects 493 Human Proteins. Suitable for cell culture supernatants. Semi-quantitative, Label-based, High Density Antibody Arrays. Sample proteins are directly conjugated to biotin, eliminating the need for a second antibody to develop the array signals. In this format, unintended antibody interactions are impossible, thus eliminating limitations on the size of the array panel. View protein list on RayBiotech’s website - AAH-BLM-2A-4
Catalog No. 50-194-8512
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.