Learn More
Raybiotech Inc L-Series Human Antibody Array 507 Glass Slide Kit.
Supplier:  Raybiotech Inc AAHBLG14
Encompass_Preferred
Detects 507 Human Proteins. Suitable for serum, plasma and cell culture supernatants. Semi-quantitative, Label-based, High Density Antibody Arrays. Sample proteins are directly conjugated to biotin, eliminating the need for a second antibody to develop the array signals. In this format, unintended antibody interactions are impossible, thus eliminating limitations on the size of the array panel. View protein list on RayBiotech’s website - AAH-BLG-1-4
Catalog No. 50-194-8491