Learn More
Sino Biological Human LAMP1 Matched ELISA Antibody Pair Set 15 Plates
Supplier:  Sino Biological SEK1121515
Encompass_Preferred
The Human LAMP-1 / CD107a / LAMP1 ELISA Pair Set is for the quantitative determination of Human LAMP-1 / CD107a / LAMP1.This ELISA Pair Set contains the basic components required for the development of sandwich ELISAs.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for LAMP-1 / CD107a / LAMP1 coated on a 96-well plate. Standards and samples are added to the wells, and any LAMP-1 / CD107a / LAMP1 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-LAMP-1 / CD107a / LAMP1 monoclonal antibody is then added, producing an antibody-antigen-antibody “sandwich”. The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of LAMP-1 / CD107a / LAMP1 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86600