Learn More
Sino Biological Human LDLR/LDL Receptor Matched ELISA Antibody Pair Set 5 Plates
Supplier:  Sino Biological SEKA102315
Encompass_Preferred
The Human LDL Receptor / LDLR ELISA Pair Set is for the quantitative determination of Human LDL Receptor / LDLR.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human LDL Receptor / LDLR coated on a 96-well plate. Standards and samples are added to the wells, and any Human LDL Receptor / LDLR present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human LDL Receptor / LDLR monoclonal antibody is then added, producing an antibody-antigen-antibody ""sandwich"". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human LDL Receptor / LDLR present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85978