Learn More
Raybiotech Inc Human Legumain ELISA Kit
Supplier:  Raybiotech Inc ELHLEGUMAIN1
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 28.7 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human 2B4 (CD244), ADAM-9, Angiopoietin-2, APRIL, BMP-2, BMP-9, C5a, Cathepsin L, CD200, CD97, Chemerin, DcR3, FABP2, FAP, FGF-19, Galectin-3, HGFR, IFN-alpha / beta R2, IGF-2, IGF-2 R, IL-1 R6 (IL-1 Rrp2), IL-24, IL-33 (IL-1 F11), Kallikrein 14.
Catalog No. 50-194-9495