Learn More
Sino Biological Recombinant Human Legumain Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 30106H08H50
Encompass_Preferred
A DNA sequence encoding the human LGMN (Q99538) (Met1-Tyr433) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-0999