Learn More
Sino Biological Human LILRA3 Matched ELISA Antibody Pair Set 15 Plates
Supplier:  Sino Biological SEK1354915
Encompass_Preferred
The Human ILT6 / LILRA3 ELISA Pair Set is for the quantitative determination of Human ILT6 / LILRA3.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human ILT6 / LILRA3 coated on a 96-well plate. Standards and samples are added to the wells, and any Human ILT6 / LILRA3 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human ILT6 / LILRA3 polyclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human ILT6 / LILRA3 present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86379