Learn More
ACROBiosystems ACROBiosystems Human LILRB3 / CD85a / ILT5 Protein, His tag
Supplier:  ACROBiosystems CDAH52H9100UG
Encompass_Preferred
Recombinant Protein;100UG;Human LILRB3, His Tag (CDA-H52H9) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - Glu 443 (Accession # AAB68668 (P288R)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 48 kDa. The protein migrates as 60-78 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6578