Learn More
Sino Biological Recombinant Anti-ILT5/LILRB3 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11978R123100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human LILRB3 (rh LILRB3; Catalog#11978-H08H; AAI04994.1; Met1-Glu443).
Catalog No. 50-241-1996