Learn More
Haematologic Technologies Inc HUMAN LYS-PLASMINOGEN 1MG
Supplier:  Haematologic Technologies Inc HCPG0133
Encompass_Preferred
Coagulation Protein, Human lys-Plasminogen, 1 milligram, MW-83,000, Research use only, 50% (v/v) glycerol/water, Store at -20 Celsius
Catalog No. 50-883-434